Anti-COX4I2 Rabbit Polyclonal Antibody
Catalog # 77704-810
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:COX4I2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q96KJ9
- Antigen synonyms:Cytochrome c oxidase subunit 4 isoform 2 mitochondrial [Precursor]|cytochrome c oxidase subunit IV isoform 2 (lung)|dJ857M17.2|COXIV 2|COX4 2|Cytochrome c oxidase subunit IV isoform 2|mitochondrial|Cytochrome c oxidase subunit 4 isoform 2|COX IV-2|RP23 106A3.3|COX4|COX42_HUMAN|COX4L2|COX4B|Cox4i2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human COX4I2
- Purification:Antigen affinity purified
- Cat. no.:77704-810
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: COX4I2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat