Anti-OR51E2 Rabbit Polyclonal Antibody
77704-670
New Product
- Antigen symbol:OR51E2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Size:100 µg
- Form:Liquid
- Gene ID:Q9H255
- Antigen synonyms:Olr59|OR51E3P|Olfactory receptor family 51 subfamily E member 2|Or51e2|Prostate-specific G-protein coupled receptor|HPRAJ|Olfactory receptor 51E2|Prostate specific G protein coupled receptor|O51E2_HUMAN|Olfactory receptor family 51 subfamily E member 3 pseudogene|Olfactory receptor OR11-16|OR52A2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human PSGR
- Purification:Antigen affinity purified
- Cat. no.:77704-670
- Packaging:Vial
Type:
Antigen: OR51E2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat