Anti-NDFIP1 Rabbit Polyclonal Antibody
Catalog # 77704-659
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:NDFIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9BT67
- Antigen synonyms:Ndfip1|Putative NFKB and MAPK-activating protein|Putative MAPK-activating protein PM13|Nedd4 family interacting protein 1|NEDD4 WW domain binding protein 5|Breast cancer associated protein SGA 1M|NEDD4 family-interacting protein 1|MGC10924|NEDD4 WW domain-binding protein 5|Breast cancer-associated protein SGA-1M|0610010M22Rik|Putative NFKB and MAPK activating protein|NFIP1_HUMAN|Putative MAPK activating protein PM13|Putative NF-kappa-B-activating protein 164|Putative NF kappa B activating protein 164|N4WBP5
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human Ndfip1
- Purification:Antigen affinity purified
- Cat. no.:77704-659
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: NDFIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat