Anti-ACAD10 Rabbit Polyclonal Antibody
77704-657
New Product
- Antigen symbol:ACAD10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q6JQN1
- Antigen synonyms:ACAD10|EC 1.3.99.-|ACAD-10|Acyl-CoA dehydrogenase family member 10
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ACAD-10
- Purification:Antigen affinity purified
- Cat. no.:77704-657
- Packaging:Vial
IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: ACAD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human