Anti-RAB11FIP1 Rabbit Polyclonal Antibody
77704-651
New Product
- Antigen symbol:RAB11FIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q6WKZ4
- Antigen synonyms:Rab11-FIP1|Rab11 FIP1|Rab-coupling protein|RCP|Rab coupling protein|RAB11 family interacting protein 1 (class I)|Rab interacting recycling protein|FLJ22524|FLJ22622|NOEL1A|No Rab11 binding protein 1A|Rab11 family-interacting protein 1|RAB11FIP1|Rab11 family interacting protein 1D|Rab effector protein|MGC78448|HGNC:30265|RFIP1_HUMAN
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human Rab11-FIP1
- Purification:Antigen affinity purified
- Cat. no.:77704-651
- Packaging:Vial
Type:
Antigen: RAB11FIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse