Anti-TUBA1A/TUBA1B/TUBA1C/TUBA3C/TUBA3E/TUBA4A Rabbit Polyclonal Antibody
77704-596
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antigen symbol:TUBA1A/TUBA1B/TUBA1C/TUBA3C/TUBA3E/TUBA4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q71U36/P68363/Q9BQE3/Q13748/Q6PEY2/P68366
- Antigen synonyms:Tubulin alpha-ubiquitous chain|TUBA6|Alpha-tubulin ubiquitous|TUBA1B|Tubulin K-alpha-1|TUBA1A|TUBA1C|Tubulin B-alpha-1|Alpha-tubulin 3|TUBA3|Tubulin alpha-1B chain|Tubulin alpha-1A chain|Tubulin alpha-3 chain|Tubulin alpha-1C c
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Tubulin α1/3/4 around the non-phosphorylation site of Y272
- Purification:Antigen affinity purified
- Cat. no.:77704-596
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: TUBA1A/TUBA1B/TUBA1C/TUBA3C/TUBA3E/TUBA4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat