Anti-PRDX2 Rabbit Polyclonal Antibody
Catalog # 77704-405
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:PRDX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P32119
- Antigen synonyms:Thioredoxin-dependent peroxide reductase 1|TPX1|PRDX2|TSA|PRX2|Torin|Thiol-specific antioxidant protein|Natural killer cell enhancing factor B|TDPX1|Peroxiredoxin-2|MGC4104|Natural Killer Enhancing Factor B|PRXII|PRDX 2|Epididymis secretory sperm binding protein Li 2a|NKEF-B|Natural killer cell-enhancing factor B|PRDX2_HUMAN|Thiol specific antioxidant protein|Thioredoxin peroxidase 1|HEL S 2a|Thioredoxin Dependent Peroxide Reductase 1|PrP|NKEF B|Thiol Specific Antioxidant 1 |Peroxiredoxin 2|NKEFB|PTX1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human PRX II
- Purification:Antigen affinity purified
- Cat. no.:77704-405
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PRDX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat