Anti-VPS72 Rabbit Polyclonal Antibody
77704-396
New Product
- Antigen symbol:VPS72
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q15906
- Antigen synonyms:Transcription factor-like 1|Protein YL-1|Swc2|VPS72|HGNC11644|Vacuolar protein sorting-associated protein 72 homolog|Vacuolar protein sorting associated protein 72 homolog|YL1|Transcription factor like 1 (YL1)|Vacuolar protein sorting 72 (yeast)|Transcription Factor Like 1|TCFL1|VPS72_HUMAN|YL-1|Vacuolar protein sorting 72 homolog (S. cerevisiae)|CFL1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human VPS72
- Purification:Antigen affinity purified
- Cat. no.:77704-396
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:40000
Type:
Antigen: VPS72
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse