Anti-TAL1 (phospho-S122) Rabbit Polyclonal Antibody
Catalog # 77704-386
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:TAL1 (phospho-S122)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P17542
- Antigen synonyms:TAL-1|Stem cell protein|TCL 5|SCL|TAL bHLH transcription factor 1 erythroid differentiation factor|Tal 1|Tal 1 product|T cell acute lymphocytic leukemia 1 protein|TAL1_HUMAN|STEM CELL LEUKEMIA HEMATOPOIETIC TRANSCRIPTION FACTOR|bHLHa17|tal1|TAL 1 protein|T cell leukemia/lymphoma 5 protein|TCL5|OTTHUMP00000009564|T-cell acute lymphocytic leukemia protein 1|Class A basic helix-loop-helix protein 17|T cell acute lymphocytic leukemia 1|T-cell leukemia/lymphoma protein 5|OTTHUMP00000009563
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human TAL1 around the phosphorylation site of S122
- Purification:Antigen affinity purified
- Cat. no.:77704-386
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TAL1 (phospho-S122)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse