Anti-TRAK2 Rabbit Polyclonal Antibody
77704-290
New Product
- Antigen symbol:TRAK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:O60296
- Antigen synonyms:Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein|OIP98|Trafficking kinesin-binding protein 2|Trak2|ALS2CR 3|GRIF1|KIAA0549|GRIF 1|Trafficking protein kinesin binding 2|Amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 3|ALS2CR3|TRAK2_HUMAN|CALS C
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human GRIF-1
- Purification:Antigen affinity purified
- Cat. no.:77704-290
- Packaging:Vial
Type:
Antigen: TRAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human