Anti-FOXD4/FOXD4L4/FOXD4L5/FOXD4L3/FOXD4L6 Rabbit Polyclonal Antibody
77704-276
New Product
- Antigen symbol:FOXD4/FOXD4L4/FOXD4L5/FOXD4L3/FOXD4L6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q12950/Q8WXT5/Q5VV16/Q6VB85/Q6VB84/Q3SYB3
- Antigen synonyms:FREAC-5|Forkhead-related transcription factor 5|Forkhead box protein D4|Forkhead box protein D4-like 4|FOXD4|FKHL9|FREAC5|FOXD4-like 4|FOXD4A|Forkhead box prote|Forkhead-related protein FKHL9|Myeloid factor-alpha|FOXD4L4|FOXD4B
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human FoxD4/D4L
- Purification:Antigen affinity purified
- Cat. no.:77704-276
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: FOXD4/FOXD4L4/FOXD4L5/FOXD4L3/FOXD4L6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human