Anti-RAF1 (phospho-S289) Rabbit Polyclonal Antibody
Catalog # 77704-036
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:RAF1 (phospho-S289)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P04049
- Antigen synonyms:OTTHUMP00000207813|v raf 1 murine leukemia viral oncogene homolog 1|v-raf murine sarcoma viral oncogene homolog 1|Raf 1|RAF1|Raf 1 proto oncogene serine/threonine kinase|RAF1_HUMAN|OTTHUMP00000160218|Craf 1 transforming gene|CMD1NN|RAF|NS5|cRaf|Murine sarcoma 3611 oncogene 1|RAF-1|C-raf|RAF proto-oncogene serine/threonine-protein kinase|EC 2.7.11.1|Oncogene MIL|Similar to murine leukemia viral (V-raf-1) oncogene homolog 1|OTTHUMP00000209389|Craf1 transforming gene|TRANSFORMING REPLICATION-DEFECTIVE MURINE RETROVIRUS 3611-MSV|v-raf-1 murine leukemia viral oncogene-like protein 1|Oncogene RAF1|vraf1 murine leukemia viral oncogene homolog 1|C-Raf proto-oncogene, serine/threonine kinase|Protein kinase raf 1|Proto-oncogene c-RAF|Raf proto oncogene serine/threonine protein kinase|kinase Raf1|c Raf
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Raf-1 around the phosphorylation site of S289
- Purification:Antigen affinity purified
- Cat. no.:77704-036
- Packaging:Vial
Specifications
About this item
Type:
Antigen: RAF1 (phospho-S289)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat