Anti-IRF1 Rabbit Polyclonal Antibody
Catalog # 77701-469
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:IRF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P10914
- Antigen synonyms:Interferon regulatory factor 1|Interferon regulatory factor 1 isoform +I9|MAR1|Interferon regulatory factor 1 isoform delta4|Interferon regulatory factor 1 isoform delta7|MAR|IRF1_HUMAN|IRF1|IRF-1|Interferon regulatory factor 1 isoform d78|IRF 1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human IRF-1
- Purification:Antigen affinity purified
- Cat. no.:77701-469
- Packaging:Vial
Specifications
About this item
Type:
Antigen: IRF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat