Anti-LAIR1 Rabbit Polyclonal Antibody
77701-446
New Product
- Antigen symbol:LAIR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q6GTX8
- Antigen synonyms:D7Bwg0421e|5133400O11Rik|LAIR 1|hLAIR1|LAIR1|LAIR1_HUMAN|Leukocyte associated Ig like receptor 1|CD305|LAIR1 protein|Leukocyte-associated immunoglobulin-like receptor 1|Leukocyte associated immunoglobulin like receptor 1|LAIR-1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CD305
- Purification:Antigen affinity purified
- Cat. no.:77701-446
- Packaging:Vial
Type:
Antigen: LAIR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human