Anti-IFNE Rabbit Polyclonal Antibody
77701-140
New Product
- Antigen symbol:IFNE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q86WN2
- Antigen synonyms:UNQ360/PRO655|Interferon epsilon-1|IFN-epsilon|IFNE|Interferon epsilon|IFNE1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human IFN-ε
- Purification:Antigen affinity purified
- Cat. no.:77701-140
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: IFNE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human