Anti-IGF2R Rabbit Polyclonal Antibody
Catalog # 77701-106
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:IGF2R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P11717
- Antigen synonyms:M6P/IGF2R|CI MPR|MPRI|300 kDa mannose 6-phosphate receptor|M6P R|CI Man-6-P receptor|300 kDa mannose 6 phosphate receptor|Insulin-like growth factor 2 receptor|MPR 300|Cation-independent mannose-6-phosphate receptor|IGF2 receptor|IGF-II receptor|MPR300|mannose 6 phosphate receptor, cation independent|CI-M6PR|M6PR|M6P/IGF2 receptor|Cation independent mannose 6 phosphate receptor|IGF 2 receptor|Igf2r|Insulin like growth factor II receptor|mannose 6 phosphate receptor|MPRI_HUMAN|IGF 2R|Insulin like growth factor 2 receptor|Insulin-like growth factor II receptor|CI-MPR|CD222 antigen|CIMPR|IGF II receptor|CD222|CI Man 6 P receptor
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human IGF-IIR
- Purification:Antigen affinity purified
- Cat. no.:77701-106
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: IGF2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human