Anti-P2RX3 Rabbit Polyclonal Antibody
Catalog # 77701-051
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:P2RX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P56373
- Antigen synonyms:Purinergic receptor P2X3|P2X receptor subunit 3|P2RX3_HUMAN|Purinergic receptor P2X ligand gated ion channel 3|Purinergic receptor|ATP receptor|P2rx3|MGC129956|P2X3|P2X purinoceptor 3|Purinoceptor P2X3
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human P2X3
- Purification:Antigen affinity purified
- Cat. no.:77701-051
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: P2RX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat