Anti-ZNF148 Rabbit Polyclonal Antibody
77700-838
New Product
- Antigen symbol:ZNF148
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9UQR1
- Antigen synonyms:Zinc finger DNA binding protein 89|ZN148_HUMAN|Transcription factor ZBP-89|ZNF148|ZBP89|Transcription factor ZBP89|Zinc finger protein 148|ZNF 148|Zinc finger DNA-binding protein 89
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human ZBP-89
- Purification:Antigen affinity purified
- Cat. no.:77700-838
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:5000
Type:
Antigen: ZNF148
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat