Anti-XRCC1 Rabbit Polyclonal Antibody
Catalog # 77700-828
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:XRCC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P18887
- Antigen synonyms:complementing defective|XRCC 1|RCC|X ray repair complementing defective repair in chinese hamster|Xrcc1|X ray repair cross complementing 1|X-ray repair cross-complementing protein 1|X ray repair cross complementing protein 1|DNA repair protein XRCC1|X ray repair|X ray Repair Complementing Defective Repair in Chinese Hamster Cells|repair in Chinese hamster|XRCC1_HUMAN|X ray repair complementing defective repair in chinese hamster cells 1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human XRCC1
- Purification:Antigen affinity purified
- Cat. no.:77700-828
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: XRCC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat