Anti-MAPT Rabbit Polyclonal Antibody
Catalog # 77700-677
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MAPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P10636
- Antigen synonyms:MGC138549|PPP1R103|FTDP 17|Microtubule associated protein tau isoform 4|MAPT|AI413597|TAU_HUMAN|TAU|Paired helical filament-tau|Protein phosphatase 1, regulatory subunit 103|MGC134287|FLJ31424|PPND|Mtapt|MAPTL|G protein beta1/gamma2 subunit interacting factor 1|MTBT1|Tauopathy and respiratory failure|Tauopathy and respiratory failure, included|MTBT2|MGC156663|PHF tau|AW045860|Microtubule-associated protein tau|Neurofibrillary tangle protein|Microtubule associated protein tau|RNPTAU|pTau|Paired helical filament tau|PHF-tau|MSTD|DDPAC
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Tau around the non-phosphorylation site of T231
- Purification:Antigen affinity purified
- Cat. no.:77700-677
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat