Anti-STK33 Rabbit Polyclonal Antibody
Catalog # 77700-642
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:STK33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9BYT3
- Antigen synonyms:STK 33|Serine/threonine kinase 33|Serine/threonine protein kinase 33|Stk33|Serine threonine kinase 33|Serine/threonine-protein kinase 33|Hypothetical protein FLJ35932|STK33_HUMAN
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human STK33
- Purification:Antigen affinity purified
- Cat. no.:77700-642
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: STK33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human