Anti-PLK2 Rabbit Polyclonal Antibody
Catalog # 77700-594
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:PLK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9NYY3
- Antigen synonyms:PLK-2|Serine/threonine-protein kinase PLK2|Serum Induced Kinase|Serum-inducible kinase|Serine/threonine protein kinase PLK2|Serine/threonine protein kinase SNK|PLK2_HUMAN|hPlk2|Polo like kinase 2|SNK|Plk2|Polo-like kinase 2|Serine/threonine-protein kinase SNK|hSNK
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Snk
- Purification:Antigen affinity purified
- Cat. no.:77700-594
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: PLK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat