Anti-INHA Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: INHA
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77707-990
- 77707-991
- Antigen symbol:
- INHA
- INHA
- Antigen synonyms:
- A inhibin subunit|A inhibin subunit precursor|Inhibin alpha subunit|INHA_HUMAN|IHA|Inhibin alpha chain precursor|inhA|Inhibin alpha chain|AW55578
- A inhibin subunit|A inhibin subunit precursor|Inhibin alpha subunit|INHA_HUMAN|IHA|Inhibin alpha chain precursor|inhA|Inhibin alpha chain|AW55578
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P05111
- P05111
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Inhibin alpha chain protein (233-366AA)
- Recombinant Human Inhibin alpha chain protein (233-366AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together