Anti-TRAF3IP3 Rabbit Polyclonal Antibody
New Product
Type:
Antigen: TRAF3IP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77707-644
- 77707-645
- Antigen symbol:
- TRAF3IP3
- TRAF3IP3
- Antigen synonyms:
- TRAF3-interacting protein 3|T3JAM|TRAF3IP3|TRAF3-interacting JNK-activating modulator
- TRAF3-interacting protein 3|T3JAM|TRAF3IP3|TRAF3-interacting JNK-activating modulator
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y228
- Q9Y228
- Isotype:
- IgG
- IgG
- Immunogen:
- Fusion protein of Human TRAF3IP3
- Fusion protein of Human TRAF3IP3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Frequently Bought Together