Anti-RARRES2 Rabbit Polyclonal Antibody
New Product
Type:
Antigen: RARRES2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77707-293
- 77707-294
- Antigen symbol:
- RARRES2
- RARRES2
- Antigen synonyms:
- Tazarotene-induced gene 2 protein|RAR-responsive protein TIG2|TIG2|RARRES2|Retinoic acid receptor responder protein 2|RARR2_HUMAN|Retinoic acid receptor responder (tazarotene induced) 2|HP10433|Tazarotene induced gene 2 protein|Retinoic acid receptor responder 2|RAR responsive protein TIG2
- Tazarotene-induced gene 2 protein|RAR-responsive protein TIG2|TIG2|RARRES2|Retinoic acid receptor responder protein 2|RARR2_HUMAN|Retinoic acid receptor responder (tazarotene induced) 2|HP10433|Tazarotene induced gene 2 protein|Retinoic acid receptor responder 2|RAR responsive protein TIG2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99969
- Q99969
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Retinoic acid receptor responder protein 2 protein (21-157AA)
- Recombinant Human Retinoic acid receptor responder protein 2 protein (21-157AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Frequently Bought Together