Anti-AGAP2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, IHC:1:50-1:200
Type:
Antigen: AGAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77707-224
- 77707-225
- Antigen symbol:
- AGAP2
- AGAP2
- Antigen synonyms:
- Cnt-g1|Centaurin gamma 1|ANK repeat and PH domains 2|ANK repeat and PH domain-containing protein 2|Agap2 ArfGAP with GTPase domain|GGAP2|GTP binding and GTPase activating protein 2|KIAA0167|Agap2|PI3K ENHANCER|Arf GAP with GTP-binding protein-like|AGAP2_HUMAN|ankyrin repeat and PH domain 2|ankyrin repeat and pleckstrin homology domains 2|GTP-binding and GTPase-activating protein 2|Phosphoinositide 3 kinase enhancer|centaurin gamma1|Nuclear GTPase PIKE|FLJ16430|Centg1|PIKE|Phosphatidylinositol 3-kinase enhancer|ArfGAP with GTPase domain|mKIAA0167|cnt g1|AGAP-2|Arf-GAP with GTPase|Cnt2|Centaurin-gamma-1|Arf GAP with GTP binding protein like|GTP-binding and GTPase activating protein 2
- Cnt-g1|Centaurin gamma 1|ANK repeat and PH domains 2|ANK repeat and PH domain-containing protein 2|Agap2 ArfGAP with GTPase domain|GGAP2|GTP binding and GTPase activating protein 2|KIAA0167|Agap2|PI3K ENHANCER|Arf GAP with GTP-binding protein-like|AGAP2_HUMAN|ankyrin repeat and PH domain 2|ankyrin repeat and pleckstrin homology domains 2|GTP-binding and GTPase-activating protein 2|Phosphoinositide 3 kinase enhancer|centaurin gamma1|Nuclear GTPase PIKE|FLJ16430|Centg1|PIKE|Phosphatidylinositol 3-kinase enhancer|ArfGAP with GTPase domain|mKIAA0167|cnt g1|AGAP-2|Arf-GAP with GTPase|Cnt2|Centaurin-gamma-1|Arf GAP with GTP binding protein like|GTP-binding and GTPase activating protein 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99490
- Q99490
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human AGAP2
- Synthetic peptide of Human AGAP2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together