Anti-MDK Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MDK
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77707-134
- 77707-135
- Antigen symbol:
- MDK
- MDK
- Antigen synonyms:
- Amphiregulin-associated protein|ARAP|MK_HUMAN|Amphiregulin associated protein|Neurite outgrowth-promoting protein|Retinoic acid inducible factor|NEGF2|MK1|NEGF 2|Neurite growth promoting factor 2|Midkine|Mdk|Midgestation and kidney protein|FLJ27379|MK 1|Neurite outgrowth-promoting factor 2|MK|Neurite outgrowth promoting protein
- Amphiregulin-associated protein|ARAP|MK_HUMAN|Amphiregulin associated protein|Neurite outgrowth-promoting protein|Retinoic acid inducible factor|NEGF2|MK1|NEGF 2|Neurite growth promoting factor 2|Midkine|Mdk|Midgestation and kidney protein|FLJ27379|MK 1|Neurite outgrowth-promoting factor 2|MK|Neurite outgrowth promoting protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21741
- P21741
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Midkine protein (21-143AA)
- Recombinant Human Midkine protein (21-143AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together