Anti-NPY1R Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100
Type:
Antigen: NPY1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Specifications
- Catalog No:
- 77707-018
- 77707-019
- Antigen symbol:
- NPY1R
- NPY1R
- Antigen synonyms:
- NPY1-R|NPYR|NPY1R|Neuropeptide Y receptor type 1|NPY1R_HUMAN|NPYY1|Neuropeptide Y receptor Y1
- NPY1-R|NPYR|NPY1R|Neuropeptide Y receptor type 1|NPY1R_HUMAN|NPYY1|Neuropeptide Y receptor Y1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25929
- P25929
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human NPY1R
- Synthetic peptide of Human NPY1R
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together