Anti-IDH1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, WB:1:200-1:1000, IHC:1:50-1:200
Type:
Antigen: IDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77720-062
- 77720-061
- Antigen symbol:
- IDH1
- IDH1
- Antigen synonyms:
- NADP dependent isocitrate dehydrogenase peroxisomal|PICD|Oxalosuccinate decarboxylase|IDCD|Isocitrate dehydrogenase [NADP] cytoplasmic|IDH|Epididymis secretory protein Li 26|ICDH|NADP dependent isocitrate dehydrogenase cytosolic|HEL-216|IDH1|IDHC_HUMAN|Cytosolic NADP-isocitrate dehydrogenase|Epididymis luminal protein 216|Isocitrate dehydrogenase 1 (NADP+) soluble|Isocitrate dehydrogenase (NADP(+)) 1 cytosolic|IDP|NADP(+)-specific ICDH|IDPC|HEL-S-26|Cytosolic NADP isocitrate dehydrogenase
- NADP dependent isocitrate dehydrogenase peroxisomal|PICD|Oxalosuccinate decarboxylase|IDCD|Isocitrate dehydrogenase [NADP] cytoplasmic|IDH|Epididymis secretory protein Li 26|ICDH|NADP dependent isocitrate dehydrogenase cytosolic|HEL-216|IDH1|IDHC_HUMAN|Cytosolic NADP-isocitrate dehydrogenase|Epididymis luminal protein 216|Isocitrate dehydrogenase 1 (NADP+) soluble|Isocitrate dehydrogenase (NADP(+)) 1 cytosolic|IDP|NADP(+)-specific ICDH|IDPC|HEL-S-26|Cytosolic NADP isocitrate dehydrogenase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75874
- O75874
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human IDH1
- Fusion protein of Human IDH1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together