Anti-PAGE1 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, WB:1:200-1:1000
Type:
Antigen: PAGE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77719-992
- 77719-993
- Antigen symbol:
- PAGE1
- PAGE1
- Antigen synonyms:
- GAGB1_HUMAN|AL5|PAGE1|GAGE 9|PAGE 1|Prostate-associated gene 1 protein|PAGE-1|G antigen family B member 1|G antigen 9|P antigen family, member 1 (prostate associated)|GAGE-9|GAGEB1|GAGE9
- GAGB1_HUMAN|AL5|PAGE1|GAGE 9|PAGE 1|Prostate-associated gene 1 protein|PAGE-1|G antigen family B member 1|G antigen 9|P antigen family, member 1 (prostate associated)|GAGE-9|GAGEB1|GAGE9
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75459
- O75459
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human PAGE1
- Fusion protein of Human PAGE1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together