Anti-CACNG8 Rabbit Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: CACNG8
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77719-967
- 77719-968
- Antigen symbol:
- CACNG8
- CACNG8
- Antigen synonyms:
- CCG8_HUMAN|TARP gamma-8|CACNG6|Calcium channel voltage dependent gamma subunit 8|Neuronal voltage gated calcium channel gamma 8 subunit|Neuronal voltage-gated calcium channel gamma-8 subunit|Voltage-dependent calcium channel gamma-8 subunit|Transmembrane AMPAR regulatory protein gamma-8|CACNG|Voltage dependent calcium channel gamma 8 subunit|Cacng8
- CCG8_HUMAN|TARP gamma-8|CACNG6|Calcium channel voltage dependent gamma subunit 8|Neuronal voltage gated calcium channel gamma 8 subunit|Neuronal voltage-gated calcium channel gamma-8 subunit|Voltage-dependent calcium channel gamma-8 subunit|Transmembrane AMPAR regulatory protein gamma-8|CACNG|Voltage dependent calcium channel gamma 8 subunit|Cacng8
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WXS5
- Q8WXS5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Peptide sequence from Human Voltage-dependent calcium channel gamma-8 subunit protein (65-83AA)
- Peptide sequence from Human Voltage-dependent calcium channel gamma-8 subunit protein (65-83AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together