Anti-STT3B Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: STT3B
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-783
- 77719-784
- Antigen symbol:
- STT3B
- STT3B
- Antigen synonyms:
- SIMP|Homolog of yeast STT3|STT3 subunit of the oligosaccharyltransferase complex homolog B|Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit STT3B|Stt3b|STT3 subunit of the oligosaccharyltransferase complex homolog B (S. cerevisiae)|Source of immunodominant MHC-associated peptides homolog|STT3-B|Oligosaccharyl transferase subunit STT3B|source of immunodominant MHC associated peptides|STT3B_HUMAN
- SIMP|Homolog of yeast STT3|STT3 subunit of the oligosaccharyltransferase complex homolog B|Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit STT3B|Stt3b|STT3 subunit of the oligosaccharyltransferase complex homolog B (S. cerevisiae)|Source of immunodominant MHC-associated peptides homolog|STT3-B|Oligosaccharyl transferase subunit STT3B|source of immunodominant MHC associated peptides|STT3B_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TCJ2
- Q8TCJ2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit STT3B protein (682-809AA)
- Recombinant Human Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit STT3B protein (682-809AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together