Anti-WDTC1 Rabbit Polyclonal Antibody
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Recommended dilution: WB:1:500-1:2000
Type:
Antigen: WDTC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Spécifications
- Catalog No:
- 77690-164
- 77719-532
- Antigen symbol:
- WDTC1
- WDTC1
- Antigen synonyms:
- WD and tetratricopeptide repeats protein 1|KIAA1037|WDTC1
- WD and tetratricopeptide repeats protein 1|KIAA1037|WDTC1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N5D0
- Q8N5D0
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human WD and tetratricopeptide repeats protein 1 protein (201-420AA)
- Recombinant Human WD and tetratricopeptide repeats protein 1 protein (201-420AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together