Anti-PIAS4 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIAS4
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-481
- 77719-482
- Antigen symbol:
- PIAS4
- PIAS4
- Antigen synonyms:
- PIAS gamma|Zinc finger MIZ type containing 6|E3 SUMO-protein ligase PIAS4|Protein inhibitor of activated STAT protein PIASy|ZMIZ6|PIAS-gamma|PIASG|PIAS 4|Protein inhibitor of activated STAT 4|MGC35296|PAIASgamma|E3 SUMO protein ligase PIAS4|PIAS4_HUMAN|Protein inhibitor of activated STAT protein 4|PIASgamma|FLJ12419|Pias4|PIASy|ZMIZ 6|Protein inhibitor of activated STAT protein gamma
- PIAS gamma|Zinc finger MIZ type containing 6|E3 SUMO-protein ligase PIAS4|Protein inhibitor of activated STAT protein PIASy|ZMIZ6|PIAS-gamma|PIASG|PIAS 4|Protein inhibitor of activated STAT 4|MGC35296|PAIASgamma|E3 SUMO protein ligase PIAS4|PIAS4_HUMAN|Protein inhibitor of activated STAT protein 4|PIASgamma|FLJ12419|Pias4|PIASy|ZMIZ 6|Protein inhibitor of activated STAT protein gamma
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N2W9
- Q8N2W9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human E3 SUMO-protein ligase PIAS4 protein (66-241AA)
- Recombinant Human E3 SUMO-protein ligase PIAS4 protein (66-241AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together