Anti-ZDHHC12 Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: ZDHHC12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77690-142
- 77719-163
- Antigen symbol:
- ZDHHC12
- ZDHHC12
- Antigen synonyms:
- ZDH12_HUMAN|Zinc finger protein 400|Zinc finger DHHC type containing 12|Zinc finger DHHC domain-containing protein 12|DHHC-12|ZNF400|ZDHHC12|Probable palmitoyltransferase ZDHHC12
- ZDH12_HUMAN|Zinc finger protein 400|Zinc finger DHHC type containing 12|Zinc finger DHHC domain-containing protein 12|DHHC-12|ZNF400|ZDHHC12|Probable palmitoyltransferase ZDHHC12
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96GR4
- Q96GR4
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Probable palmitoyltransferase ZDHHC12 protein
- Recombinant Human Probable palmitoyltransferase ZDHHC12 protein
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together