Anti-FKBP10 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FKBP10
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-039
- 77719-040
- Antigen symbol:
- FKBP10
- FKBP10
- Antigen synonyms:
- FKBP-65|Immunophilin FKBP65|FKBP65|65 kDa FK506-binding protein|PSEC0056Peptidyl-prolyl cis-trans isomerase FKBP10|FKBP-10|FKBP10|EC 5.2.1.8|Rotamase|65 kDa FKBP|FK506-binding protein 10|PPIase FKBP10
- FKBP-65|Immunophilin FKBP65|FKBP65|65 kDa FK506-binding protein|PSEC0056Peptidyl-prolyl cis-trans isomerase FKBP10|FKBP-10|FKBP10|EC 5.2.1.8|Rotamase|65 kDa FKBP|FK506-binding protein 10|PPIase FKBP10
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96AY3
- Q96AY3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Peptidyl-prolyl cis-trans isomerase FKBP10 protein (459-579AA)
- Recombinant Human Peptidyl-prolyl cis-trans isomerase FKBP10 protein (459-579AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together