Anti-ND-75 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ND-75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila melanogaster
Specifications
- Catalog No:
- 77718-962
- 77718-963
- Antigen symbol:
- ND-75
- ND-75
- Antigen synonyms:
- ND75|ND-75|Complex I-75kD|mitochondrial|CG2286|EC 7.1.1.2|EC 1.6.99.3|CI-75kD|NADH-ubiquinone oxidoreductase 75 kDa subunit
- ND75|ND-75|Complex I-75kD|mitochondrial|CG2286|EC 7.1.1.2|EC 1.6.99.3|CI-75kD|NADH-ubiquinone oxidoreductase 75 kDa subunit
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Drosophila melanogaster
- Drosophila melanogaster
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q94511
- Q94511
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Drosophila melanogaster NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial protein (28-315AA)
- Recombinant Drosophila melanogaster NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial protein (28-315AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together