Anti-RORB Rabbit Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: RORB
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77718-925
- 77718-926
- Antigen symbol:
- RORB
- RORB
- Antigen synonyms:
- Nuclear receptor subfamily 1 group F member 2|Retinoic acid binding receptor beta|Nuclear receptor ROR-beta|Retinoid-related orphan receptor-beta|NR1F2|Nuclear receptor RZR beta|RAR related orphan receptor B|RAR related orphan receptor beta|RZRB|Rorb|Nuclear receptor RZR-beta|ROR BETA|OTTHUMP00000021486|RZR BETA|RORB_HUMAN|bA133M9.1
- Nuclear receptor subfamily 1 group F member 2|Retinoic acid binding receptor beta|Nuclear receptor ROR-beta|Retinoid-related orphan receptor-beta|NR1F2|Nuclear receptor RZR beta|RAR related orphan receptor B|RAR related orphan receptor beta|RZRB|Rorb|Nuclear receptor RZR-beta|ROR BETA|OTTHUMP00000021486|RZR BETA|RORB_HUMAN|bA133M9.1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92753
- Q92753
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Nuclear receptor ROR-beta protein (1-269AA)
- Recombinant Human Nuclear receptor ROR-beta protein (1-269AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together