Order Entry
Canada
ContactUsLinkComponent
Anti-CPT1B Rabbit Polyclonal Antibody (FITC)
Anti-CPT1B Rabbit Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-CPT1B Rabbit Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77718-884
  • 77718-883
  • Antigen symbol:
  • CPT1B
  • CPT1B
  • Antigen synonyms:
  • FLJ58750|CPTIM|CPT1 M|M CPT1|KIAA1670|Carnitine palmitoyltransferase 1B (muscle)|CPTI|Carnitine O palmitoyltransferase I muscle isoform|FLJ55729|Carnitine O palmitoyltransferase I mitochondrial muscle isoform|CPTI-M|CPT1 muscle|M-CPT1|CPT 1B|Carnitine O-palmitoyltransferase I|Carnitine O palmitoyltransferase 1B|Cpt1b|CPT1B_HUMAN|CPT I|CPTI M|Carnitine palmitoyltransferase 1B|Carnitine O-palmitoyltransferase 1|CPT1-M|Carnitine palmitoyltransferase I muscle|Carnitine palmitoyltransferase I like protein|muscle isoform|MCCPT1|CPTI muscle|CPT1M|Carnitine palmitoyltransferase I-like protein|MCPT1|Carnitine palmitoyltransferase 1A (muscle)
  • FLJ58750|CPTIM|CPT1 M|M CPT1|KIAA1670|Carnitine palmitoyltransferase 1B (muscle)|CPTI|Carnitine O palmitoyltransferase I muscle isoform|FLJ55729|Carnitine O palmitoyltransferase I mitochondrial muscle isoform|CPTI-M|CPT1 muscle|M-CPT1|CPT 1B|Carnitine O-palmitoyltransferase I|Carnitine O palmitoyltransferase 1B|Cpt1b|CPT1B_HUMAN|CPT I|CPTI M|Carnitine palmitoyltransferase 1B|Carnitine O-palmitoyltransferase 1|CPT1-M|Carnitine palmitoyltransferase I muscle|Carnitine palmitoyltransferase I like protein|muscle isoform|MCCPT1|CPTI muscle|CPT1M|Carnitine palmitoyltransferase I-like protein|MCPT1|Carnitine palmitoyltransferase 1A (muscle)
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q92523
  • Q92523
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Carnitine O-palmitoyltransferase 1, muscle isoform protein (165-318AA)
  • Recombinant Human Carnitine O-palmitoyltransferase 1, muscle isoform protein (165-318AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together