Anti-TPH2 (phospho-Ser19) Rabbit Polyclonal Antibody
Catalog # 77708-608
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:TPH2 (phospho-Ser19)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µl
- Form:Liquid
- Gene ID:Q8IWU9
- Antigen synonyms:MGC138872|Tryptophan hydroxylase 2|Tryptophan 5-monooxygenase 2|TPH2_HUMAN|TPH 2|FLJ37295|MGC138871|Tph2|Tryptophan 5-hydroxylase 2|NTPH|Neuronal tryptophan hydroxylase|ADHD7
- Storage buffer:PBS, 150bmM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of Serine 19(G-F-S(p)-L-D) derived from Human TPH2
- Purification:Antigen affinity purified
- Cat. no.:77708-608
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TPH2 (phospho-Ser19)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat