Anti-AKAP1 Rabbit Polyclonal Antibody
Catalog # 77706-625
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:AKAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q92667
- Antigen synonyms:DAKAP1|Dual specificity A-kinase-anchoring protein 1|Protein kinase A-anchoring protein 1|D-AKAP-1|Protein kinase A1|mitochondrial|AKAP 149|AKAP1_HUMAN|A-kinase anchor protein 1|AKAP|Akap1|SAKAP84|Spermatid A-kinase anchor protein 84|S-AKAP84|PRKA1|A kinase anchor protein|A-kinase anchor protein 149 kDa
- Storage buffer:PBS, 150bmM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human AKAP1
- Purification:Antigen affinity purified
- Cat. no.:77706-625
- Packaging:Vial
Specifications
About this item
WB:1:500-1:3000, IHC:1:50-1:100, IF:1:100-1:500
Type:
Antigen: AKAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human