Anti-ARNT2 Rabbit Polyclonal Antibody
77705-147
New Product
- Antigen symbol:ARNT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9HBZ2
- Antigen synonyms:Aryl hydrocarbon receptor nuclear translocator 2|ARNT2_HUMAN|ARNT protein 2|bHLHe1|ARNT2|Class E basic helix-loop-helix protein 1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human Arnt 2
- Purification:Antigen affinity purified
- Cat. no.:77705-147
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: ARNT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat