Anti-TBC1D4 (phospho-T642) Rabbit Polyclonal Antibody
Catalog # 77705-141
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:TBC1D4 (phospho-T642)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:O60343
- Antigen synonyms:Acrg embryonic lethality (mouse) minimal region ortholog|Acrg embryonic lethality minimal region ortholog|AS160|Akt substrate of 160 kDa|TBC (Tre 2 BUB2 CDC16) domain containing protein|TBC1 domain family member 4|CDC16|AS 160|TBCD4_HUMAN|TBC1 D4|Acrg embryonic lethality mouse minimal region ortholog|NIDDM5|KIAA0603|TBC Tre 2 BUB2 CDC16 domain containing protein|Tre-2|BUB2|Tbc1d4
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human TBC1D4 around the phosphorylation site of T642
- Purification:Antigen affinity purified
- Cat. no.:77705-141
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TBC1D4 (phospho-T642)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse