Anti-CCKAR Rabbit Polyclonal Antibody
Catalog # 77704-922
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CCKAR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P32238
- Antigen synonyms:CCKA receptor|Cholecystokinin-1 receptor|Cholecystokinin type A receptor|Cholecystokinin A receptor|CCK1-R|CCK-A receptor|Cholecystokinin 1 receptor|Cholecystokinin receptor type A|CCKAR_HUMAN|CCK-AR|CCKAR|CCK AR|CCK A|CCKR Type A|CCK A receptor|CCKRA|CCKA|CCK1 R|CCK1R
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CCK-AR
- Purification:Antigen affinity purified
- Cat. no.:77704-922
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: CCKAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat