Anti-RUNX1 (phospho-S276) Rabbit Polyclonal Antibody
77704-866
New Product
- Antigen symbol:RUNX1 (phospho-S276)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q01196
- Antigen synonyms:CBFA 2|Oncogene AML 1|OTTHUMP00000108700|PEBP2aB|CBFA2|SL3-3 enhancer factor 1 alpha B subunit|alpha subunit core binding factor|SL3/AKV core binding factor alpha B subunit|Runt-related transcription factor 1|Run1|OTTHUMP00000108696|AMLCR1|PEA2-alpha B|SL3/AKV core-binding factor alpha B subunit|OTTHUMP00000108699|RUNX 1|AML1 EVI 1 fusion protein|PEBP2-alpha B|AML1 EVI 1|SL3 3 enhancer factor 1 alpha B subunit|Polyomavirus enhancer binding protein 2 alpha B subunit|Aml1 oncogene|RUNX1_HUMAN|PEBP2 alpha B|Core-binding factor subunit alpha-2|Polyomavirus enhancer-binding protein 2 alpha B subunit|PEBP2A2|Runt related transcription factor 1|Runx1|PEA2 alpha B|EVI 1|CBF alpha 2|HGNC|AML1|Core binding factor runt domain alpha subunit 2|Oncogene AML-1|Core binding factor alpha 2 subunit|AMLCR 1|CBF-alpha-2|Acute myeloid leukemia 1|EVI1|Acute myeloid leukemia 1 protein|OTTHUMP00000108702|OTTHUMP00000108697|AML 1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human RUNX1 around the phosphorylation site of S276
- Purification:Antigen affinity purified
- Cat. no.:77704-866
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: RUNX1 (phospho-S276)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat