Anti-ACTL9 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ACTL9
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-145
- 77722-146
- Antigen symbol:
- ACTL9
- ACTL9
- Antigen synonyms:
- hypothetical protein MGC33407|Actl9|hypothetical protein LOC284382|ACTL7C|HSD21|actin-like 9|Actin-like protein 9|ACTL9_HUMAN
- hypothetical protein MGC33407|Actl9|hypothetical protein LOC284382|ACTL7C|HSD21|actin-like 9|Actin-like protein 9|ACTL9_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TC94
- Q8TC94
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Actin-like protein 9 protein (1-160AA)
- Recombinant Human Actin-like protein 9 protein (1-160AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together