Anti-MYRIP Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MYRIP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-107
- 77722-108
- Antigen symbol:
- MYRIP
- MYRIP
- Antigen synonyms:
- Slp homolog lacking C2 domains c|Exophilin 8|Exophilin8|DKFZp586F1018|MYRIP_HUMAN|Myosin VIIA and Rab interacting protein|Slp homologue lacking C2 domains|Exophilin|Myrip|MGC130034|Slp homologue lacking C2 domains c|Exophilin-8|SLAC2 C|SlaC2-c|Synaptotagmin like protein homologue lacking C2 domains c|Myosin-VIIa- and Rab-interacting protein|SLAC2C|MGC130035|SLAC 2C|Rab effector MyRIP|synaptotagmin-like protein lacking C2 domains C|FLJ44025
- Slp homolog lacking C2 domains c|Exophilin 8|Exophilin8|DKFZp586F1018|MYRIP_HUMAN|Myosin VIIA and Rab interacting protein|Slp homologue lacking C2 domains|Exophilin|Myrip|MGC130034|Slp homologue lacking C2 domains c|Exophilin-8|SLAC2 C|SlaC2-c|Synaptotagmin like protein homologue lacking C2 domains c|Myosin-VIIa- and Rab-interacting protein|SLAC2C|MGC130035|SLAC 2C|Rab effector MyRIP|synaptotagmin-like protein lacking C2 domains C|FLJ44025
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NFW9
- Q8NFW9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Rab effector MyRIP protein (387-689AA)
- Recombinant Human Rab effector MyRIP protein (387-689AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together