Order Entry
Canada
ContactUsLinkComponent
Anti-CISD2 Rabbit Polyclonal Antibody
Anti-CISD2 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-CISD2 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77690-274
  • 77721-992
  • Antigen symbol:
  • CISD2
  • CISD2
  • Antigen synonyms:
  • B630006A20Rik|Zcd2|CISD2|1500031D15Rik|Endoplasmic reticulum intermembrane small protein|1500026J14Rik|CDGSH iron-sulfur domain-containing protein 2|Nutrient deprivation autophagy factor 1|AI848398|NAF-1|Zinc finger|WFS2|Noxp70|ERIS|RGD1566242|CDGSH iron sulfur domain 2|MitoNEET-related 1 protein|CDGSH type domain 2|OTTHUMP00000219576|1500009M05Rik|Nutrient-deprivation autophagy factor-1|CISD2_HUMAN|MitoNEET related 1|Miner1
  • B630006A20Rik|Zcd2|CISD2|1500031D15Rik|Endoplasmic reticulum intermembrane small protein|1500026J14Rik|CDGSH iron-sulfur domain-containing protein 2|Nutrient deprivation autophagy factor 1|AI848398|NAF-1|Zinc finger|WFS2|Noxp70|ERIS|RGD1566242|CDGSH iron sulfur domain 2|MitoNEET-related 1 protein|CDGSH type domain 2|OTTHUMP00000219576|1500009M05Rik|Nutrient-deprivation autophagy factor-1|CISD2_HUMAN|MitoNEET related 1|Miner1
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q8N5K1
  • Q8N5K1
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human CDGSH iron-sulfur domain-containing protein 2 protein (61-135AA)
  • Recombinant Human CDGSH iron-sulfur domain-containing protein 2 protein (61-135AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together