Anti-VSIG2 Rabbit Polyclonal Antibody
New Product
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: VSIG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77721-738
- 77721-737
- Antigen symbol:
- VSIG2
- VSIG2
- Antigen synonyms:
- 2210413P10Rik|Ctm|VSIG2_HUMAN|V set and immunoglobulin domain containing 2|CTXL|CTH|V-set and immunoglobulin domain-containing protein 2|VSIG2|Cortical thymocyte-like protein|cortical thymocyte receptor (X. laevis CTX) like|CT-like protein|OTTHUMP00000197171|OTTHUMP00000197172
- 2210413P10Rik|Ctm|VSIG2_HUMAN|V set and immunoglobulin domain containing 2|CTXL|CTH|V-set and immunoglobulin domain-containing protein 2|VSIG2|Cortical thymocyte-like protein|cortical thymocyte receptor (X. laevis CTX) like|CT-like protein|OTTHUMP00000197171|OTTHUMP00000197172
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96IQ7
- Q96IQ7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human V-set and immunoglobulin domain-containing protein 2 protein (24-243AA)
- Recombinant Human V-set and immunoglobulin domain-containing protein 2 protein (24-243AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together