Anti-SLC35C1 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: SLC35C1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77721-578
- 77721-579
- Antigen symbol:
- SLC35C1
- SLC35C1
- Antigen synonyms:
- Solute carrier family 35 member C1|FUCT1GDP-fucose transporter 1|SLC35C1
- Solute carrier family 35 member C1|FUCT1GDP-fucose transporter 1|SLC35C1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96A29
- Q96A29
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human GDP-fucose transporter 1 protein (287-364AA)
- Recombinant Human GDP-fucose transporter 1 protein (287-364AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together